Sodium methanesulfonate ≥98.0%

Supplier: TCI America

Synonyms: Methanesulfonic acid sodium salt, Methanesulphonic acid sodium salt

S0518-25G S0518-500G
TCS0518-25GEA 67.94 CAD
TCS0518-25G TCS0518-500G
Sodium methanesulfonate ≥98.0%
Sodium methanesulphonate

CAS Number: 2386-57-4
Molecular Formula: CH4O3S
Molecular Weight: 118.08
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White


Other Product Names / Synonyms: Methanesulfonic Acid Sodium Salt

Formula: CH₃NaO₃S
MDL Number: MFCD00064389
CAS Number: 2386-57-4
EINECS: 219-203-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2383cc654c8e884067f1d5998cf17055e3e6612e&vid=2383cc654c8e884067f1d5998cf17055e3e6612e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR