Methyl linoleate ≥98.0%

Supplier: TCI America

Synonyms: cis,cis-9,12-Octadecadienoic acid methyl ester, 9,12-Octadecanoic acid methyl ester, 9Z,12Z-Octadecadienoic acid methyl ester, Linoleic acid methyl ester, (9Z,12Z)-Methyl octadeca-9,12-dienoate, Methyl cis,cis-9,12-octadecadienoate, 9Z,12Z-Oktadekadiensäuremethylester, 9,12 Methyl octadecaotenoate methyl linoleate, cis,cis-9,12-Oktadekadiensäuremethylester

S0325-5G S0325-1G
TCS0325-005GEA 139.7 CAD
TCS0325-005G TCS0325-001G
Methyl linoleate ≥98.0%
Methyl linoleate

[Standard Material for GC]
CAS Number: 112-63-0
MDL Number: MFCD00009534
Molecular Formula: C19H34O2
Molecular Weight: 294.48
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 346
Melting point (°C): -35
Flash Point (°C): 200
Specific Gravity (20/20): 0.89
Storage Temperature: 0-10°C


Product Categories: Bioscience, Higher Fatty Acids & Higher Alcohols, Analytical Science, Fatty Acid Methyl Esters (GC Standard)
Other Product Names / Synonyms: Linoleic Acid Methyl Ester

Formula: C₁₉H₃₄O₂
MW: 294.48 g/mol
Boiling Pt: 340
Melting Pt: -35
Density: 0.89 (20 °C)
Flash Pt: 200 °C
MDL Number: MFCD00009534
CAS Number: 112-63-0
EINECS: 203-993-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6b25e42b08c1484d386c720347209afa52607f57&vid=50481ef163a2a9251e1fc41ae00335a25d67274d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR