n-Tetradecane ≥99.5%

Supplier: TCI America

Synonyms: Tetradecane

S0286-5ML
TCS0286-005MLEA 80.58 CAD
TCS0286-005ML
n-Tetradecane ≥99.5%
n-Tetradecane

[Standard Material for GC]
CAS Number: 629-59-4
MDL Number: MFCD00008986
Molecular Formula: C14H30
Molecular Weight: 198.39
Purity/Analysis Method: >99.5% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 253
Melting point (°C): 6
Flash Point (°C): 100
Specific Gravity (20/20): 0.76

Formula: H₃C(CH₂)₁₂CH₃
MW: 198.39 g/mol
Boiling Pt: 253 °C
Melting Pt: 6 °C
Density: 0.76 (20 °C)
Flash Pt: 100 °C
MDL Number: MFCD00008986
CAS Number: 629-59-4
https://ca.vwr.com/stibo/web/std.lang.all/86/19/738619.jpg

Order Now

Specification Test Results

Purity/Analysis Method 99.5% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR