Sudan III ≥90.0% (by UV method)

Supplier: TCI America

S0142-25G S0142-100G
TCS0142-025GEA 42.66 CAD
TCS0142-025G TCS0142-100G
Sudan III ≥90.0% (by UV method)
Sudan III

CAS Number: 85-86-9
MDL Number: MFCD00003905
Molecular Formula: C22H16N4O
Molecular Weight: 352.40
Purity/Analysis Method: >90.0% (E)
Form: Crystal
Color: Yellow Red
Melting point (°C): 202
Lambda max.: 513 nm (CHCl3)


Product Categories: Bioscience, Stains & Dyes
Other Product Names / Synonyms: Oil Red AS, Solvent Red 23

Formula: C₂₂H₁₆N₄O
Melting Pt: 202 °C
MDL Number: MFCD00003905
CAS Number: 85-86-9
EINECS: 201-638-4
Merck Index: 12,09054
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=652a7f66bb70072b318c95c7c4a71c94cfc7b26b&vid=652a7f66bb70072b318c95c7c4a71c94cfc7b26b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 90.0% (E)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR