Docusate sodium ≥95.0% (by titrimetric analysis)

Supplier: TCI America
Warning

Synonyms: Dioctyl sulphosuccinate sodium salt, Dioctyl sulphosuccinate, sodium salt, Sodium dioctyl sulphosuccinate, Dioctyl sulfosuccinate sodium salt, Sodium 1,4-bis((2-ethylhexyl)oxy)-1,4-dioxobutane-2-sulphonate, bis(2-Ethylhexyl) sodium sulphosuccinate, Sodium 1,4-bis((2-ethylhexyl)oxy)-1,4-dioxobutane-2-sulfonate, Sodium dioctyl sulfosuccinate, Dioctyl sulfosuccinate, sodium salt, bis(2-Ethylhexyl) sodium sulfosuccinate

S0139-25G S0139-500G
TCS0139-025GEA 40.2 CAD
TCS0139-025G TCS0139-500G
Docusate sodium ≥95.0% (by titrimetric analysis)
Docusate sodium

CAS Number: 577-11-7
MDL Number: MFCD00012455
Molecular Formula: C20H38O7S
Molecular Weight: 444.56
Purity/Analysis Method: >95.0% (T)
Form: Spongy Solid
Color: White
Melting point (°C): 179


Product Categories: Materials Chemistry, Anionic Surfactants
Other Product Names / Synonyms: Dioctyl Sulfosuccinate Sodium Salt, Sulfosuccinic Acid Bis(2-ethylhexyl) Ester Sodium Salt, Docusate Sodium Salt

Formula: C₂₀H₃₇NaO₇S
MW: 444.57 g/mol
Boiling Pt: ∼180 °C (1013 hPa)
Melting Pt: 179 °C
Density: ∼1.1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00012455
CAS Number: 577-11-7
EINECS: 209-406-4
Merck Index: 12,03460
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9c8f290e0c10bff9d169923f0343d666b014c8a6&vid=6814085cc168cf417f66157cfc59a04bab5aaa9e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR