Sulfanilic acid ≥99.0%

Supplier: TCI America
Warning

Synonyms: Aniline-4-sulphonic acid, 4-Aminobenzenesulphonic acid, 4-Aminobenzenesulfonic acid, Aniline-4-sulfonic acid

S0120-25G S0120-500G
TCS0120-025GEA 58.86 CAD
TCS0120-025G TCS0120-500G
Sulfanilic acid ≥99.0%
Sulphanilic acid

CAS Number: 121-57-3
MDL Number: MFCD00007886
Molecular Formula: C6H7NO3S
Molecular Weight: 173.19
Purity/Analysis Method: >99.0% (HPLC,T)
Form: Crystal
Color: White
Melting point (°C): 288


Product Categories: Bioscience, ROS/RNS Detection Reagents, Reagents for Nitric Oxide Research
Other Product Names / Synonyms: 4-Aminobenzenesulfonic Acid, p-Anilinesulfonic Acid, 4-Sulfoaniline

Formula: 4-(H₂N)C₆H₄SO₃H
Boiling Pt: 363 °C (1013 hPa)
Melting Pt: 288 °C
Density: 1.485 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007886
CAS Number: 121-57-3
EINECS: 204-482-5
Merck Index: 13,09012
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b06ceb5c3db7565cdc317b2d31e0c8e34e4863a4&vid=b06ceb5c3db7565cdc317b2d31e0c8e34e4863a4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR