Ammonium sulfamate ≥98.5%

Supplier: TCI America
Warning

Synonyms: Ammonium sulfamidate, Ammonium sulphamidate, Ammonium amidosulfonate, Ammonium amidosulphonate

S0117-500G S0117-25G
TCS0117-500GEA 164.87 CAD
TCS0117-500G TCS0117-25G
Ammonium sulfamate ≥98.5%
Ammonium sulphamate

CAS Number: 7773-06-0
MDL Number: MFCD00011429
Molecular Formula: H3NO3S
Molecular Weight: 114.12
Purity/Analysis Method: >98.5% (T)
Form: Crystal
Color: White


Other Product Names / Synonyms: Aminosulfonic Acid Ammonium Salt, Ammonium Sulfamate, Sulfamic Acid Ammonium Salt

Formula: H₆N₂O₃S
MW: 114.13 g/mol
Melting Pt: 131.1
Density: 1 g/cm³ (20 °C)
MDL Number: MFCD00011429
CAS Number: 7773-06-0
EINECS: 231-871-7
Merck Index: 12,00589
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=59ea2306572e26653ba7676750fb84daf6b6bdba&vid=59ea2306572e26653ba7676750fb84daf6b6bdba&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.5% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR