D-(+)-Sucrose ≥99.0%

Supplier: TCI America

Synonyms: β-ᴅ-Fructofuranosyl-α-ᴅ-glucopyranoside, α-ᴅ-Glucopyranosyl β-ᴅ-fructofuranoside, D(+)-Saccharose, α-ᴅ-Glc-(1→2)-β-ᴅ-Fru, Sucrose

S0111-25G S0111-500G
TCS0111-25GEA 35.38 CAD
TCS0111-25G TCS0111-500G
D-(+)-Sucrose ≥99.0%
D(+)-Sucrose

CAS Number: 57-50-1
MDL Number: MFCD00006626
Molecular Formula: C12H22O11
Molecular Weight: 342.30
Purity/Analysis Method: >99.0% (HPLC)
Form: Crystal
Color: White
Melting point (°C): 187
Specific rotation [a]20/D: 66.5 deg (C=26, H2O)


Product Categories: Bioscience, Basic Sugars (Mono & Oligosaccharides), Disaccharides
Other Product Names / Synonyms: D-(+)-Saccharose

Formula: C₁₂H₂₂O₁₁
MW: 342.3 g/mol
Melting Pt: 187 °C
Density: 1.29 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006626
CAS Number: 57-50-1
EINECS: 200-334-9
Merck Index: 13,08966
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=038a5006987727fe38dba215db032d061146434c&vid=038a5006987727fe38dba215db032d061146434c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR