Succinic anhydride 95,0

Supplier: TCI America
Warning

Synonyms: Dihydrofuran-2,5-dione

S0107-25G S0107-500G
TCS0107-25GEA 38.63 CAD
TCS0107-25G TCS0107-500G
Succinic anhydride 95,0
Succinic anhydride

CAS Number: 108-30-5
MDL Number: MFCD00005525
Molecular Formula: C4H4O3
Molecular Weight: 100.07
Purity/Analysis Method: >95.0% (T)
Form: Pellet
Color: White
Boiling point (°C): 131
Melting point (°C): 120
Flash Point (°C): 148


Product Categories: Bioscience, Functionality Creating Reagents
Other Product Names / Synonyms: Dihydro-2,5-furandione

Formula: C₄H₄O₃
MW: 100.07 g/mol
Boiling Pt: 261
Melting Pt: 120
Density: 1.503 g/cm³ (20 °C)
Flash Pt: 148 °C
MDL Number: MFCD00005525
CAS Number: 108-30-5
EINECS: 203-570-0
UN: 3261
ADR: 8 III
Merck Index: 12,09039
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=39e57e8ffde94c95b2c601da550c362d9782e7c1&vid=39e57e8ffde94c95b2c601da550c362d9782e7c1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR