Dimethyl succinate ≥98.0%
Supplier: TCI America
|
Warning
|
S0104-25G
S0104-500G
TCS0104-025GEA
38.57
CAD
TCS0104-025G
TCS0104-500G
Dimethyl succinate ≥98.0%
Dimethyl succinate
CAS Number: 106-65-0
MDL Number: MFCD00008466
Molecular Formula: C6H10O4
Molecular Weight: 146.14
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 196
Flash Point (°C): 98
Freezing point (°C): 18
Specific Gravity (20/20): 1.12
Product Categories: Materials Chemistry, Resin Additives
Other Product Names / Synonyms: Succinic Acid Dimethyl Ester, Methyl Succinate
Formula:
C₆H₁₀O₄ Boiling Pt: 196 °C Melting Pt: 18…19 °C Density: 1.12 (20 °C) Flash Pt: 98 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008466 CAS Number: 106-65-0 EINECS: 203-419-9 Merck Index: 12,09038 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...