Diethyl succinate ≥99.0%

Supplier: TCI America

S0103-25G S0103-500G
TCS0103-025GEA 36.89 CAD
TCS0103-025G TCS0103-500G
Diethyl succinate ≥99.0%
Diethyl succinate

CAS Number: 123-25-1
MDL Number: MFCD00009208
Molecular Formula: C8H14O4
Molecular Weight: 174.20
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 196
Flash Point (°C): 104
Specific Gravity (20/20): 1.04


Product Categories: Materials Chemistry, Resin Additives
Other Product Names / Synonyms: Succinic Acid Diethyl Ester, Ethyl Succinate

Formula: C₈H₁₄O₄
Boiling Pt: 196 °C
Melting Pt: –19 °C
Density: 1.04 (20 °C)
Flash Pt: 104 °C
Storage Temperature: Ambient
MDL Number: MFCD00009208
CAS Number: 123-25-1
EINECS: 204-612-0
Merck Index: 12,09038
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ab1ef2ddf5f88e954d3e53dc48a2988020b2dd55&vid=ab1ef2ddf5f88e954d3e53dc48a2988020b2dd55&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR