Succinic acid ≥99.0%

Supplier: TCI America
Warning

Synonyms: 1,4-Butanedioic acid

S0100-500G S0100-25G
TCS0100-500GEA 57.84 CAD
TCS0100-500G TCS0100-025G
Succinic acid ≥99.0%
Succinic acid

CAS Number: 110-15-6
MDL Number: MFCD00002789
Molecular Formula: C4H6O4
Molecular Weight: 118.09
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Color: White
Boiling point (°C): 235
Melting point (°C): 187
Flash Point (°C): 206


Other Product Names / Synonyms: Butanedioic Acid

Formula: HOOCCH₂CH₂COOH
MW: 118.09 g/mol
Boiling Pt: 235
Melting Pt: 182
Density: 1.56 g/cm³ (20 °C)
Flash Pt: 206
Storage Temperature: Ambient
MDL Number: MFCD00002789
CAS Number: 110-15-6
EINECS: 203-740-4
UN: 3261
ADR: 8 III
Merck Index: 13,08953
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9b4744e028c2dd66fc6553370daff69dea95225c&vid=9b4744e028c2dd66fc6553370daff69dea95225c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR