Succinamide ≥98.0% (by titrimetric analysis)

Supplier: TCI America

S0096-25G S0096-500G
TCS0096-025GEA 80.86 CAD
TCS0096-025G TCS0096-500G
Succinamide ≥98.0% (by titrimetric analysis)
Succinamide

CAS Number: 110-14-5
MDL Number: MFCD00008042
Molecular Formula: C4H8N2O2
Molecular Weight: 116.12
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White

Formula: C₄H₈N₂O₂
MDL Number: MFCD00008042
CAS Number: 110-14-5
EINECS: 203-739-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=123f19f41edad2558df42f84e0bb2f0c211cb818&vid=123f19f41edad2558df42f84e0bb2f0c211cb818&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR