trans-Stilbene ≥98.0%

Supplier: TCI America

Synonyms: trans-1,2-Diphenylethylene, (E)-1,2-Diphenylethene, trans-1,2-Diphenylethene

S0090-25G S0090-100G
TCS0090-025GEA 199.08 CAD
TCS0090-025G TCS0090-100G
trans-Stilbene ≥98.0%
trans-Stilbene

CAS Number: 103-30-0
MDL Number: MFCD00064300
Molecular Formula: C14H12
Molecular Weight: 180.25
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 307
Melting point (°C): 123


Product Categories: Analytical Science, Scintillators
Other Product Names / Synonyms: trans-1,2-Diphenylethylene

Formula: C₁₄H₁₂
MW: 180.25 g/mol
Boiling Pt: 307 °C
Melting Pt: 123 °C
Density: 0.9707 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064300
CAS Number: 103-30-0
EINECS: 203-098-5
Merck Index: 12,08972
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0332af8e0e5f881798d621790bb996f5f535c1f8&vid=0332af8e0e5f881798d621790bb996f5f535c1f8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR