cis-Stilbene ≥96.0%

Supplier: TCI America

Synonyms: cis-1,2-Diphenylethene, Isostilbene, cis-1,2-Diphenylethylene, (Z)-1,2-Diphenylethene

S0089-5ML S0089-1ML
TCS0089-005MLEA 129.56 CAD
TCS0089-005ML TCS0089-1ML
cis-Stilbene ≥96.0%
cis-Stilbene

CAS Number: 645-49-8
MDL Number: MFCD00004788
Molecular Formula: C14H12
Molecular Weight: 180.25
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Color: Very Pale Yellow
Boiling point (°C): 152
Flash Point (°C): 110
Specific Gravity (20/20): 1.02


Other Product Names / Synonyms: cis-1,2-Diphenylethylene

Formula: (C₆H₅)CHCH(C₆H₅)
Boiling Pt: 152 °C
Density: 1.02 (20 °C)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00004788
CAS Number: 645-49-8
EINECS: 211-445-7
Merck Index: 12,08972
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c0a968ffbca7aced95ca6de5610cf523fd064c8d&vid=c0a968ffbca7aced95ca6de5610cf523fd064c8d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR