Ethyl stearate ≥96.0%
Supplier: TCI America
S0079-25G
S0079-250G
TCS0079-025GEA
62.71
CAD
TCS0079-025G
TCS0079-250G
Ethyl stearate ≥96.0%
Ethyl stearate
CAS Number: 111-61-5
MDL Number: MFCD00009006
Molecular Formula: C20H40O2
Molecular Weight: 312.54
Purity/Analysis Method: >96.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 199
Flash Point (°C): 110
Freezing point (°C): 30
Product Categories: Bioscience, Lipids
Other Product Names / Synonyms: Stearic Acid Ethyl Ester
Formula:
C₂₀H₄₀O₂ Boiling Pt: 199 °C Melting Pt: 31 °C Density: 0.8973 g/cm³ (25 °C) Flash Pt: 110 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00009006 CAS Number: 111-61-5 EINECS: 203-887-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...