D-(-)-Sorbitol ≥97.0%

Supplier: TCI America

Synonyms: (2R,3R,4R,5S)-Hexane-1,2,3,4,5,6-hexaol, (Sorbitol), D(-)-Glucitol, D-Sorbitol, Sorbitol

S0065-25G S0065-500G
TCS0065-25GEA 40.4 CAD
TCS0065-25G TCS0065-500G
D-(-)-Sorbitol ≥97.0%
D(-)-Sorbitol

CAS Number: 50-70-4
MDL Number: MFCD00004708
Molecular Formula: C6H14O6
Molecular Weight: 182.17
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 295
Melting point (°C): 100
Specific rotation [a]20/D: 106 deg (C=0.4, Acidified (NH4)6Mo7O24)

Formula: HOH₂C(CH(OH))₄CH₂OH
MW: 182.17 g/mol
Boiling Pt: 295
Melting Pt: 100 °C
Density: 1.524 g/cm³ (20 °C)
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00004708
CAS Number: 50-70-4
EINECS: 200-061-5
Merck Index: 13,08797
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0723a28bc08683d1b0acc20d82babb4a39125057&vid=0723a28bc08683d1b0acc20d82babb4a39125057&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR