Span® 60

Supplier: TCI America

S0062-25G S0062-500G
TCS0062-025GEA 44.11 CAD
TCS0062-025G TCS0062-500G
Span® 60
Span® 60

CAS Number: 1338-41-6
MDL Number: MFCD00005366
Form: Crystal
Color: Very Pale Yellow
Melting point (°C): 52
Flash Point (°C): 232


Other Product Names / Synonyms: Arlacel 60, Sorbitan Monostearate

Formula: C₂₄H₄₆O₆
Boiling Pt: ∼100 °C (1013 hPa)
Melting Pt: 52 °C
Density: 1 g/cm³ (20 °C)
Flash Pt: 232 °C
Storage Temperature: Ambient
MDL Number: MFCD00005366
CAS Number: 1338-41-6
EINECS: 215-664-9
Merck Index: 12,08872

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR