m-Cresol purple sodium salt

Supplier: TCI America

Synonyms: m-Cresolsulfonphthalein sodium salt, m-Cresolsulphonphthalein sodium salt, 3,3-Bis(4-hydroxy-2-methylphenyl)-3H-benzo[d][1,2]oxathiole 2,2-dioxide, sodium salt

S0045-1G S0045-5G
TCS0045-001GEA 67.94 CAD
TCS0045-001G TCS0045-5G
m-Cresol purple sodium salt
m-Cresol purple sodium salt

CAS Number: 62625-31-4
MDL Number: MFCD00010177
Molecular Formula: C21H18O5S
Molecular Weight: 404.41
Form: Crystal
Color: Deep Yellow Red


Other Product Names / Synonyms: m-Cresol Purple Sodium Salt, Sodium m-Cresol Purple, Sodium Metacresol Purple

Formula: C₂₁H₁₇NaO₅S
MW: 404.42 g/mol
MDL Number: MFCD00010177
CAS Number: 62625-31-4
EINECS: 263-656-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=19ac168a98e4d1760efb8f5297c7fb43006d01cc&vid=19ac168a98e4d1760efb8f5297c7fb43006d01cc&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR