Bromothymol blue sodium salt

Supplier: TCI America

S0044-25G S0044-5G
TCS0044-25GEA 408.35 CAD
TCS0044-25G TCS0044-005G
Bromothymol blue sodium salt
Bromothymol blue sodium salt

CAS Number: 34722-90-2
MDL Number: MFCD00077263
Molecular Formula: C27H28Br2O5S
Molecular Weight: 646.37
Form: Crystal
Color: Reddish Yellow


Other Product Names / Synonyms: Sodium Bromothymol Blue

Formula: C₂₇H₂₇Br₂NaO₅S
Melting Pt: > 200 °C
Storage Temperature: Ambient
MDL Number: MFCD00077263
CAS Number: 34722-90-2
EINECS: 252-169-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0f69fed2cbfe6ad641d2c9d79336f3b541c12b17&vid=0f69fed2cbfe6ad641d2c9d79336f3b541c12b17&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR