Bromophenol blue sodium salt

Supplier: TCI America

S0043-5G S0043-1G
TCS0043-5GEA 228.03 CAD
TCS0043-5G TCS0043-001G
Bromophenol blue sodium salt
Bromophenol blue sodium salt

CAS Number: 34725-61-6
MDL Number: MFCD00013793
Molecular Formula: C19H10Br4O5S
Molecular Weight: 691.94
Form: Crystal
Color: Deep Green


Other Product Names / Synonyms: Sodium Bromophenol Blue, BPB

Formula: C₁₉H₉Br₄NaO₅S
Storage Temperature: Ambient
MDL Number: MFCD00013793
CAS Number: 34725-61-6
EINECS: 252-170-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=631bb19c87f709c5c36309aa854f6125fce19663&vid=631bb19c87f709c5c36309aa854f6125fce19663&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR