Bromocresol green sodium salt ≥95.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: Bromocresol green water soluble, Sodium 2,6-dibromo-4-(3-(3,5-dibromo-4-hydroxy-2-methylphenyl)-2,2-dioxido-3H-benzo[d][1,2]oxathiol-3-yl)-3-methylphenolate

S0041-5G S0041-1G
TCS0041-5GEA 650.96 CAD
TCS0041-5G TCS0041-001G
Bromocresol green sodium salt ≥95.0% (by HPLC, titration analysis)
Bromocresol green sodium salt

CAS Number: 1007478-52-5
MDL Number: MFCD00148898
Molecular Formula: C21H14Br4O5S
Molecular Weight: 720.00
Purity/Analysis Method: >95.0% (HPLC,T)
Form: Crystal
Color: Deep Yellow Red
Melting point (°C): 230
Lambda max.: 612 nm


Product Categories: Analytical Science, Indicators (pH)
Other Product Names / Synonyms: Bromocresol Blue Sodium Salt, Sodium Bromocresol Green

Formula: C₂₁H₁₃Br₄NaO₅S
MW: 720 g/mol
Melting Pt: 230 °C
Storage Temperature: Ambient
MDL Number: MFCD00148898
CAS Number: 62625-32-5
EINECS: 263-657-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=112766df65da8f45bbf53ac09d49b5986362776c&vid=112766df65da8f45bbf53ac09d49b5986362776c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR