(-)-Shikimic acid ≥97.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: (3R,4S,5R)-3,4,5-Trihydroxycyclohex-1-ene-1-carboxylic acid, Shikimic acid

S0038-100MG S0038-1G S0038-5G
TCS0038-100MGEA 37.92 CAD
TCS0038-100MG TCS0038-001G TCS0038-5G
(-)-Shikimic acid ≥97.0% (by titrimetric analysis)
(-)-Shikimic acid

CAS Number: 138-59-0
MDL Number: MFCD00066278
Molecular Formula: C7H10O5
Molecular Weight: 174.15
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Color: White
Melting point (°C): 185
Specific rotation [a]20/D: -180 deg (C=1, H2O)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (3R,4S,5R)-(-)-3,4,5-Trihydroxy-1-cyclohexene-1-carboxylic Acid

Formula: C₇H₁₀O₅
MW: 174.15 g/mol
Melting Pt: 185 °C
MDL Number: MFCD00066278
CAS Number: 138-59-0
EINECS: 205-334-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e06266cdfbd5e7c04bcf60068ac4f6b152e6fcb4&vid=e06266cdfbd5e7c04bcf60068ac4f6b152e6fcb4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR