L(+)-Serine ≥99.0%

Supplier: TCI America

Synonyms: H-Ser-OH, (S)-2-Amino-3-hydroxypropanoic acid, Serine, (S)-(+)-Serine

S0035-25G S0035-250G S0035-5G
TCS0035-025GEA 66.04 CAD
TCS0035-025G TCS0035-250G TCS0035-5G
L(+)-Serine ≥99.0%
L(+)-Serine

CAS Number: 56-45-1
MDL Number: MFCD00064224
Molecular Formula: C3H7NO3
Molecular Weight: 105.09
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Color: White
Melting point (°C): 228
Specific rotation [a]20/D: 15 deg (C=10, HCl(1+5))


Product Categories: Bioscience, Proteinogenic Amino Acids
Other Product Names / Synonyms: L-2-Amino-3-hydroxypropionic Acid, H-Ser-OH

Formula: C₃H₇NO₃
MW: 105.09 g/mol
Boiling Pt: 259
Melting Pt: 228 °C
Density: 1.6 g/cm³ (22 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064224
CAS Number: 56-45-1
EINECS: 200-274-3
Merck Index: 14,00846
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=07dc9e7e2067940ae1ee9b24ba0b00de02f28b94&vid=07dc9e7e2067940ae1ee9b24ba0b00de02f28b94&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR