DL-Serine ≥98.0%

Supplier: TCI America

Synonyms: (±)-Serine, H-DL-Ser-OH, DL-2-Amino-3-hydroxypropionic acid

S0034-500G S0034-25G
TCS0034-500GEA 229.91 CAD
TCS0034-500G TCS0034-025G
DL-Serine ≥98.0%
DL-Serine

CAS Number: 302-84-1
MDL Number: MFCD00064223
Molecular Formula: C3H7NO3
Molecular Weight: 105.09
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Melting point (°C): 240


Product Categories: Bioscience, Proteinogenic Amino Acids
Other Product Names / Synonyms: DL-2-Amino-3-hydroxypropionic Acid, H-DL-Ser-OH

Formula: C₃H₇NO₃
MW: 105.09 g/mol
Boiling Pt: 395
Melting Pt: 240 °C
Density: 1.543 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064223
CAS Number: 302-84-1
EINECS: 206-130-6
Merck Index: 13,08534
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ab5addbc470f3eca473fa1b3a1d1cae3f1c20ad&vid=4ab5addbc470f3eca473fa1b3a1d1cae3f1c20ad&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR