D-(-)-Serine ≥98.5%

Supplier: TCI America

Synonyms: (R)-(-)-Serine, H-D-Ser-OH

S0033-25G S0033-5G
TCS0033-025GEA 131.14 CAD
TCS0033-025G TCS0033-5G
D-(-)-Serine ≥98.5%
D(-)-Serine

CAS Number: 312-84-5
MDL Number: MFCD00004269
Molecular Formula: C3H7NO3
Molecular Weight: 105.09
Purity/Analysis Method: >98.5% (T)
Form: Crystal
Color: White
Specific rotation [a]20/D: -15 deg (C=10, HCl(1+5))


Product Categories: Bioscience, Proteinogenic Amino Acids
Other Product Names / Synonyms: D-2-Amino-3-hydroxypropionic Acid, H-D-Ser-OH

Formula: C₃H₇NO₃
MW: 105.09 g/mol
Melting Pt: 235
MDL Number: MFCD00004269
CAS Number: 312-84-5
EINECS: 206-229-4
Merck Index: 12,08604
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dad51a80202c1a334c9909fe58b339861731a9d1&vid=dad51a80202c1a334c9909fe58b339861731a9d1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.5% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR