Dimethyl sebacate ≥98.0%

Supplier: TCI America

Synonyms: 1,10-Decanedioic acid dimethyl ester, Dimethyl decanedioate, Sebacic acid dimethyl ester

S0027-25G S0027-500G
TCS0027-025GEA 47.59 CAD
TCS0027-025G TCS0027-500G
Dimethyl sebacate ≥98.0%
Dimethyl sebacate

CAS Number: 106-79-6
MDL Number: MFCD00008472
Molecular Formula: C12H22O4
Molecular Weight: 230.30
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 158
Flash Point (°C): 145
Freezing point (°C): 26


Product Categories: Materials Chemistry, Resin Additives
Other Product Names / Synonyms: Decanedioic Acid Dimethyl Ester, Sebacic Acid Dimethyl Ester, Dimethyl Decanedioate

Formula: C₁₂H₂₂O₄
MW: 230.3 g/mol
Boiling Pt: 102
Melting Pt: 38
Density: 1.0989 g/cm³ (20 °C)
Flash Pt: 145 °C
Storage Temperature: Ambient
MDL Number: MFCD00008472
CAS Number: 106-79-6
EINECS: 203-431-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c827258cdb842ba71b0bfde1d801517a3b52f1be&vid=c827258cdb842ba71b0bfde1d801517a3b52f1be&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR