Dibutyl sebacate ≥98.0%
Supplier: TCI America
|
Warning
|
Synonyms:
Sebacic acid dibutyl ester, 1,10-Decanedioic acid dibutyl ester, Dibutyl decanedioate
CAS Number: 109-43-3
MDL Number: MFCD00027218
Molecular Formula: C18H34O4
Molecular Weight: 314.47
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 345
Melting point (°C): -10
Flash Point (°C): 190
Specific Gravity (20/20): 0.94
Product Categories: Materials Chemistry, Resin Additives
Other Product Names / Synonyms: Decanedioic Acid Dibutyl Ester, Dibutyl Decanedioate, Sebacic Acid Dibutyl Ester
Formula:
C₁₈H₃₄O₄ MW: 314.47 g/mol Boiling Pt: 345 °C Melting Pt: –10 °C Density: 0.94 (20 °C) Flash Pt: 190 °C Storage Temperature: Ambient |
MDL Number:
MFCD00027218 CAS Number: 109-43-3 EINECS: 203-672-5 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...