Sebacic acid ≥98.0%

Supplier: TCI America

Synonyms: Decanedioic acid, 1,8-Octanedicarboxylic acid, 1,10-Decanedioic acid

S0022-25G S0022-500G
TCS0022-025GEA 36.47 CAD
TCS0022-025G TCS0022-500G
Sebacic acid ≥98.0%
Sebacic acid

CAS Number: 111-20-6
MDL Number: MFCD00004440
Molecular Formula: C10H18O4
Molecular Weight: 202.25
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: White
Boiling point (°C): 295
Melting point (°C): 134
Flash Point (°C): 219


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: Decanedioic Acid, 1,8-Octanedicarboxylic Acid

Formula: C₁₀H₁₈O₄
MW: 202.25 g/mol
Boiling Pt: 374
Melting Pt: 134 °C
Density: 1.21 g/cm³ (20 °C)
Flash Pt: 219 °C
Storage Temperature: Ambient
MDL Number: MFCD00004440
CAS Number: 111-20-6
EINECS: 203-845-5
REACH: 01-2119519212-52
Merck Index: 12,08558
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=45e845d41668215b673f8feb5d7dcff16c5f8047&vid=45e845d41668215b673f8feb5d7dcff16c5f8047&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR