Saponin

Supplier: TCI America

S0019-250G S0019-25G
TCS0019-250GEA 1128.97 CAD
TCS0019-250G TCS0019-025G
Saponin
Saponin

CAS Number: 8047-15-2
MDL Number: MFCD00081981
Form: Crystal
Color: Very Pale Yellow


Product Categories: Bioscience, Sugars (Others), Glycosides

Density: 1.015…1.02 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00081981
CAS Number: 8047-15-2
EINECS: 232-462-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR