Phenyl salicylate ≥98.0%

Supplier: TCI America
Warning

Synonyms: Phenyl 2-hydroxybenzoate, Phenyl Salicylate

S0017-25G S0017-500G
TCS0017-025GEA 42.66 CAD
TCS0017-025G TCS0017-500G
Phenyl salicylate ≥98.0%
Phenyl salicylate

CAS Number: 118-55-8
MDL Number: MFCD00002213
Molecular Formula: C13H10O3
Molecular Weight: 214.22
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 173
Melting point (°C): 44
Flash Point (°C): 113


Other Product Names / Synonyms: 2-Hydroxybenzoic Acid Phenyl Ester, Phenyl 2-Hydroxybenzoate, Salicylic Acid Phenyl Ester

Formula: C₁₃H₁₀O₃
MW: 214.22 g/mol
Boiling Pt: 307
Melting Pt: 44 °C
Density: 1.307 g/cm³ (20 °C)
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00002213
CAS Number: 118-55-8
EINECS: 204-259-2
UN: 3077
ADR: 9 III
Merck Index: 12,07464
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d190f2c627160ab825dd3186ff7725875aa0ee48&vid=d190f2c627160ab825dd3186ff7725875aa0ee48&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR