Benzyl salicylate ≥99.0%
Supplier: TCI America
Synonyms:
Benzyl 2-hydroxybenzoate
S0009-25G
S0009-500G
TCS0009-025GEA
36.34
CAD
TCS0009-025G
TCS0009-500G
Benzyl salicylate ≥99.0%
Benzyl salicylate
CAS Number: 118-58-1
MDL Number: MFCD00020034
Molecular Formula: C14H12O3
Molecular Weight: 228.25
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 194
Flash Point (°C): 184
Specific Gravity (20/20): 1.18
Other Product Names / Synonyms: Salicylic Acid Benzyl Ester, 2-Hydroxybenzoic Acid Benzyl Ester, Benzyl 2-Hydroxybenzoate
Formula:
C₁₄H₁₂O₃ MW: 228.25 g/mol Boiling Pt: 208 Melting Pt: 23.9 Density: 1.18 (20 °C) Flash Pt: 184 °C |
MDL Number:
MFCD00020034 CAS Number: 118-58-1 EINECS: 204-262-9 Merck Index: 13,01146 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...