Salicylamide ≥98.0% (by titrimetric analysis)
Supplier: TCI America
|
Warning
|
S0006-25G
S0006-500G
TCS0006-025GEA
33.39
CAD
TCS0006-025G
TCS0006-500G
Salicylamide ≥98.0% (by titrimetric analysis)
Salicylamide
CAS Number: 65-45-2
MDL Number: MFCD00007978
Molecular Formula: C7H7NO2
Molecular Weight: 137.14
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Boiling point (°C): 182
Melting point (°C): 140
Other Product Names / Synonyms: 2-Hydroxybenzamide
Formula:
C₇H₇NO₂ Boiling Pt: 182 °C Melting Pt: 140 °C Density: 1.33 g/cm³ (20 °C) |
MDL Number:
MFCD00007978 CAS Number: 65-45-2 EINECS: 200-609-3 Merck Index: 12,08480 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...