Salicylaldoxime ≥98.0%

Supplier: TCI America

Synonyms: 2-Hydroxybenzaldehyde oxime

S0005-25G S0005-500G
TCS0005-025GEA 75.57 CAD
TCS0005-025G TCS0005-500G
Salicylaldoxime ≥98.0%
Salicylaldoxime

CAS Number: 94-67-7
MDL Number: MFCD00002120
Molecular Formula: C7H7NO2
Molecular Weight: 137.14
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 57

Formula: C₇H₇NO₂
Melting Pt: 57 °C
MDL Number: MFCD00002120
CAS Number: 94-67-7
EINECS: 202-353-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9b9b6abfe8c05c98aab75298ed29401c72a53048&vid=9b9b6abfe8c05c98aab75298ed29401c72a53048&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR