L(+)-Ribose ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: (2S,3S,4S)-2,3,4,5-Tetrahydroxypentanal, L-(+)-Ribose

R0068-5G R0068-1G R0068-25G
TCR0068-5GEA 64.31 CAD
TCR0068-5G TCR0068-001G TCR0068-25G
L(+)-Ribose ≥98.0% (by HPLC)
L(+)-Ribose

CAS Number: 24259-59-4
MDL Number: MFCD00167010
Molecular Formula: C5H10O5
Molecular Weight: 150.13
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Color: White
Melting point (°C): 90
Specific rotation [a]20/D: 20 deg (C=1, H2O)

Formula: C₅H₁₀O₅
MW: 150.13 g/mol
Boiling Pt: 331 °C (1013 hPa)
Melting Pt: 90 °C
Storage Temperature: Ambient
MDL Number: MFCD00167010
CAS Number: 24259-59-4
EINECS: 246-110-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=85ccbedbd80447375505bc2c4d35ba21bfadf597&vid=85ccbedbd80447375505bc2c4d35ba21bfadf597&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR