Rhodanine-N-acetic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America

R0054-25G R0054-5G
TCR0054-025GEA 82.16 CAD
TCR0054-025G TCR0054-5G
Rhodanine-N-acetic acid ≥98.0% (by titrimetric analysis)
Rhodanine-N-acetic acid

CAS Number: 5718-83-2
MDL Number: MFCD00005491
Molecular Formula: C5H5NO3S2
Molecular Weight: 191.22
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: Very Pale Yellow
Melting point (°C): 147


Other Product Names / Synonyms: N-Carboxymethylrhodanine

Formula: C₅H₅NO₃S₂
Melting Pt: 147 °C
MDL Number: MFCD00005491
CAS Number: 5718-83-2
EINECS: 227-220-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=faa755606ecdd76c3e32891bb8129f89dc6ea3c2&vid=faa755606ecdd76c3e32891bb8129f89dc6ea3c2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR