1,3-Diacetoxybenzene ≥98.0%
Supplier: TCI America
R0046-25G
R0046-500G
TCR0046-025GEA
44.24
CAD
TCR0046-025G
TCR0046-500G
1,3-Diacetoxybenzene ≥98.0%
1,3-Diacetoxybenzene
CAS Number: 108-58-7
MDL Number: MFCD00008701
Molecular Formula: C10H10O4
Molecular Weight: 194.19
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 278
Flash Point (°C): 110
Specific Gravity (20/20): 1.18
Other Product Names / Synonyms: 1,3-Phenylene Diacetate, Resorcinol Diacetate
Formula:
C₁₀H₁₀O₄ Boiling Pt: 278 °C Melting Pt: 142…143 °C Density: 1.18 (20 °C) Flash Pt: 110 °C |
MDL Number:
MFCD00008701 CAS Number: 108-58-7 EINECS: 203-596-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...