D-(-)-Ribose ≥98.0%

Supplier: TCI America

R0025-250G R0025-25G
TCR0025-250GEA 170.96 CAD
TCR0025-250G TCR0025-025G
D-(-)-Ribose ≥98.0%
D(-)-Ribose

CAS Number: 50-69-1
MDL Number: MFCD00135453
Molecular Formula: C5H10O5
Molecular Weight: 150.13
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Specific rotation [a]20/D: -21 deg (C=1, H2O)

Formula: C₅H₁₀O₅
MW: 150.13 g/mol
Boiling Pt: 331 °C (1013 hPa)
Melting Pt: 90…95 °C
Density: 0.8 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00135453
CAS Number: 50-69-1
EINECS: 200-059-4
Merck Index: 12,08371
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=47fb4068d14650f4f7e6b4d08c25bc54826877a2&vid=47fb4068d14650f4f7e6b4d08c25bc54826877a2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR