Riboflavine (vitamin B2) ≥98%

Supplier: TCI America

Synonyms: Vitamin B2, Lactoflavin, 7,8-Dimethyl-10-((2S,3S,4R)-2,3,4,5-tetrahydroxypentyl)benzo[g]pteridine-2,4(3H,10H)-dione, (-)-Riboflavin, Vitamine G

R0020-25G R0020-500G R0020-100G
TCR0020-025GEA 89.44 CAD
TCR0020-025G TCR0020-500G TCR0020-100G
Riboflavine (vitamin B2) ≥98%
Riboflavine (Vitamin B2)

CAS Number: 83-88-5
MDL Number: MFCD00005022
Molecular Formula: C17H20N4O6
Molecular Weight: 376.37
Form: Crystal
Color: Yellow
Specific rotation [a]20/D: -135 deg (C=0.5, JP Method)


Other Product Names / Synonyms: Lactoflavin, Vitamin B2

Formula: C₁₇H₂₀N₄O₆
MW: 376.37 g/mol
Melting Pt: 280 °C
Storage Temperature: Ambient
MDL Number: MFCD00005022
CAS Number: 83-88-5
EINECS: 201-507-1
Merck Index: 12,08367
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7bb57aac1397f17d62dbf6ca57ea637e40c75bba&vid=7bb57aac1397f17d62dbf6ca57ea637e40c75bba&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR