Rhodizonic acid disodium salt ≥90.0% (by titrimetric analysis)

Supplier: TCI America

R0019-25G R0019-1G
TCR0019-025GEA 696.15 CAD
TCR0019-025G TCR0019-001G
Rhodizonic acid disodium salt ≥90.0% (by titrimetric analysis)
Rhodizonic acid disodium salt

CAS Number: 523-21-7
MDL Number: MFCD00001650
Molecular Formula: C6H2O6
Molecular Weight: 214.04
Purity/Analysis Method: >90.0% (T)
Form: Crystal
Color: Yellow
Melting point (°C): 300


Other Product Names / Synonyms: Rhodizonic Acid Disodium Salt, Sodium Rhodizonate, 3,4,5,6-Tetraoxocyclohexene-1,2-diol Disodium Salt

Formula: C₆Na₂O₆
Melting Pt: 300 °C
MDL Number: MFCD00001650
CAS Number: 523-21-7
EINECS: 208-340-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=14c5b65f7d7b74fc6c1f10e0f36d0a995185fe27&vid=14c5b65f7d7b74fc6c1f10e0f36d0a995185fe27&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 90.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR