4-Quinolinecarbaldehyde ≥97.0%
Supplier: TCI America
Synonyms:
Quinoline-4-carbaldehyde, Quinoline-4-carboxaldehyde, 4-Quinolinecarboxaldehyde
Q0061-1G
Q0061-5G
Q0061-25G
TCQ0061-001GEA
61.62
CAD
TCQ0061-001G
TCQ0061-005G
TCQ0061-25G
4-Quinolinecarbaldehyde ≥97.0%
4-Quinolinecarbaldehyde
CAS Number: 4363-93-3
MDL Number: MFCD00006781
Molecular Formula: C10H7NO
Molecular Weight: 157.17
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Color: Very Pale Yellow
Boiling point (°C): 123
Melting point (°C): 50
Flash Point (°C): 110
Other Product Names / Synonyms: Cinchoninaldehyde, 4-Formylquinoline
Formula:
C₁₀H₇NO Boiling Pt: 123 °C Melting Pt: 50 °C Flash Pt: 110 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00006781 CAS Number: 4363-93-3 EINECS: 224-453-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...