5-Quinolinol ≥98.0%

Supplier: TCI America
Warning

Synonyms: 5-Hydroxyquinoline, Quinolin-5-ol

Q0059-1G Q0059-5G
TCQ0059-001GEA 79 CAD
TCQ0059-001G TCQ0059-005G
5-Quinolinol ≥98.0%
5-Quinolinol

CAS Number: 578-67-6
MDL Number: MFCD00006792
Molecular Formula: C9H7NO
Molecular Weight: 145.16
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: Pale Yellow


Other Product Names / Synonyms: 5-Hydroxyquinoline

Formula: C₉H₇NO
MW: 145.16 g/mol
Boiling Pt: 313
Melting Pt: 223…226 °C
Density: 1.57 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006792
CAS Number: 578-67-6
EINECS: 209-428-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a5045dd4cc5ea33f11f796306669504ccc32d7eb&vid=8b92cd77c8d9347962ff15cf146134e8e7ed48cf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR