3-Quinuclidinone hydrochloride ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 1-Azabicyclo[2.2.2]octan-3-one hydrochloride

Q0052-5G Q0052-25G
TCQ0052-005GEA 34.76 CAD
TCQ0052-005G TCQ0052-25G
3-Quinuclidinone hydrochloride ≥98.0% (by titrimetric analysis)
3-Quinuclidinone hydrochloride

CAS Number: 1193-65-3
MDL Number: MFCD00137391
Molecular Formula: C7H11NO
Molecular Weight: 161.63
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Melting point (°C): 300


Other Product Names / Synonyms: 1-Azabicyclo[2.2.2]octan-3-one Hydrochloride

Formula: C₇H₁₂ClNO
Melting Pt: 300 °C
MDL Number: MFCD00137391
CAS Number: 1193-65-3
EINECS: 214-776-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c2822e60c67a97dace89fd239cda9c7b0e79a71d&vid=c2822e60c67a97dace89fd239cda9c7b0e79a71d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR