4-Quinolinol ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Q0029-5G Q0029-25G
TCQ0029-005GEA 101.12 CAD
TCQ0029-005G TCQ0029-25G
4-Quinolinol ≥98.0% (by GC, titration analysis)
4-Quinolinol

CAS Number: 611-36-9
MDL Number: MFCD00006777
Molecular Formula: C9H7NO
Molecular Weight: 145.16
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: White
Melting point (°C): 203


Other Product Names / Synonyms: 4-Hydroxyquinoline

Formula: C₉H₇NO
Melting Pt: 203 °C
Storage Temperature: Ambient
MDL Number: MFCD00006777
CAS Number: 611-36-9
EINECS: 210-268-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4151d5f4ec692c1b4fc0d65d8b6b71df21360019&vid=54e0ac0748b1d5a2c2d6e7af321d3ad5a369611d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR