D-(-)-Quinic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Q0009-25G Q0009-5G
TCQ0009-025GEA 231.45 CAD
TCQ0009-025G TCQ0009-5G
D-(-)-Quinic acid ≥98.0% (by titrimetric analysis)
D(-)-Quinic acid

CAS Number: 77-95-2
MDL Number: MFCD00003864
Molecular Formula: C7H12O6
Molecular Weight: 192.17
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Melting point (°C): 166
Specific rotation [a]20/D: -43.5 deg (C=10, H2O)


Product Categories: Synthetic Organic Chemistry, Optical Resolving Agents, Chiral Building Blocks
Other Product Names / Synonyms: D-(-)-Chinic Acid, D-(-)-Kinic Acid, 1,3,4,5-Tetrahydroxycyclohexanecarboxylic Acid

Formula: C₇H₁₂O₆
Melting Pt: 166 °C
MDL Number: MFCD00003864
CAS Number: 77-95-2
EINECS: 201-072-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=276b52e9cbc9636342d541b2b94c9bc87a5ad2b1&vid=276b52e9cbc9636342d541b2b94c9bc87a5ad2b1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR