L(+)-Potassium sodium tartrate tetrahydrate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Potassium sodium tartrate tetrahydrate, Potassium sodium (2R,3R)-2,3-dihydroxysuccinate tetrahydrate, Potassium sodium L(+)-tartrate tetrahydrate, Sodium potassium tartrate, (R,R)-(+)-Tartaric acid potassium sodium salt tetrahydrate, (R,R)-(+)-Potassium sodium tartrate tetrahydrate, Rochelle salt, L(+)-Tartaric acid potassium sodium salt tetrahydrate, (+)-Potassium sodium L-tartrate tetrahydrate

P1798-25G P1798-500G P1798-100G
TCP1798-25GEA 48.98 CAD
TCP1798-25G TCP1798-500G TCP1798-100G
L(+)-Potassium sodium tartrate tetrahydrate ≥98.0% (by titrimetric analysis)
L(+)-Potassium sodium tartrate tetrahydrate

CAS Number: 6381-59-5
MDL Number: MFCD00150989
Molecular Formula: C4H6O6
Molecular Weight: 210.16
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Melting point (°C): 80
Specific rotation [a]20/D: 22 deg (C=10, H2O)


Other Product Names / Synonyms: L-(+)-Tartaric Acid Potassium Sodium Salt, Rochelle Salt, Seignette Salt

Formula: KOOC(CHOH)₂COONa·4H₂O
MW: 282.22 g/mol
Melting Pt: 80 °C
Density: ∼1.767 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00150989
CAS Number: 6381-59-5
EINECS: 206-156-8
Merck Index: 13,07755
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=34213bc48026e8ee327641c6b32ef3655bc1ea3d&vid=34213bc48026e8ee327641c6b32ef3655bc1ea3d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR