1,4-Phenyldiboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America

Synonyms: p-Phenylenebisboronic acid, 1,4-Phenylenediboronic acid, 1,4-Phenyldiboric acid, 1,4-Benzenediboronic acid

P1358-1G P1358-5G P1358-25G
TCP1358-001GEA 52.14 CAD
TCP1358-001G TCP1358-5G TCP1358-25G
1,4-Phenyldiboronic acid (contains varying amounts of Anhydride)
1,4-Phenyldiboronic acid

CAS Number: 4612-26-4
MDL Number: MFCD00236018
Molecular Formula: C6H8B2O4
Molecular Weight: 165.75
Form: Crystal
Color: White


Other Product Names / Synonyms: 1,4-Benzenediboronic Acid

Formula: C₆H₈B₂O₄
MDL Number: MFCD00236018
CAS Number: 4612-26-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c139fa33642eecd1301d7ae3ad80bbf6e47739db&vid=c139fa33642eecd1301d7ae3ad80bbf6e47739db&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR