(R)-(+)-2-Pyrrolidinone-5-carboxylic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: D-Pyr-OH, (R)-(+)-2-Pyrrolidone-5-carboxylic acid

P1354-25G P1354-5G
TCP1354-025GEA 265.44 CAD
TCP1354-025G TCP1354-005G
(R)-(+)-2-Pyrrolidinone-5-carboxylic acid ≥98.0% (by titrimetric analysis)
(R)-(+)-2-Pyrrolidinone-5-carboxylic acid

CAS Number: 4042-36-8
MDL Number: MFCD00066212
Molecular Formula: C5H7NO3
Molecular Weight: 129.12
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Melting point (°C): 162
Specific rotation [a]20/D: 10 deg (C=5, H2O)


Product Categories: Synthetic Organic Chemistry, Optical Resolving Agents, Bioscience, Biologically-modified Amino Acids
Other Product Names / Synonyms: D-Glutamic Acid Lactam, (R)-5-Oxopyrrolidine-2-carboxylic Acid, (R)-2-Pyrrolidone-5-carboxylic Acid, H-D-Pyr-OH

Formula: C₅H₇NO₃
MW: 129.12 g/mol
Melting Pt: 162 °C
MDL Number: MFCD00066212
CAS Number: 4042-36-8
EINECS: 223-735-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=926a56527b9dc5df86df98f056aa1f2356d6ce7e&vid=926a56527b9dc5df86df98f056aa1f2356d6ce7e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR