Dihexyl phthalate ≥98.0%

Supplier: TCI America

P1344-25G P1344-5G
TCP1344-25GEA 239.56 CAD
TCP1344-25G TCP1344-005G
Dihexyl phthalate ≥98.0%
Dihexyl phthalate

CAS Number: 84-75-3
MDL Number: MFCD00043720
Molecular Formula: C20H30O4
Molecular Weight: 334.46
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 333
Flash Point (°C): 177
Specific Gravity (20/20): 1.01


Product Categories: Materials Chemistry, Resin Additives, Analytical Science, Phthalates (Environmental Endocrine Disruptors)
Other Product Names / Synonyms: Phthalic Acid Dihexyl Ester

Formula: C₂₀H₃₀O₄
MW: 334.46 g/mol
Boiling Pt: 333 °C
Density: 1.01 (20 °C)
Flash Pt: 177 °C
MDL Number: MFCD00043720
CAS Number: 84-75-3
EINECS: 201-559-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3f13aa1c535f7f563505214464215f86d14b13f8&vid=3f13aa1c535f7f563505214464215f86d14b13f8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR