3-Phenoxy-1,2-propandiol ≥95.0%
Supplier: TCI America
P1335-5G
P1335-25G
TCP1335-005GEA
189.6
CAD
TCP1335-005G
TCP1335-025G
3-Phenoxy-1,2-propandiol ≥95.0%
3-Phenoxy-1,2-propandiol
CAS Number: 538-43-2
MDL Number: MFCD00004717
Molecular Formula: C9H12O3
Molecular Weight: 168.19
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 202
Melting point (°C): 54
Flash Point (°C): 113
Other Product Names / Synonyms: Glycerol alpha-Monophenyl Ether
Formula:
C₆H₅OCH₂CH(OH)CH₂OH Boiling Pt: 202 °C Melting Pt: 54 °C Density: 1.1477 g/cm³ (20 °C) Flash Pt: 113 °C |
MDL Number:
MFCD00004717 CAS Number: 538-43-2 EINECS: 208-692-8 Merck Index: 12,07444 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...